Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSK3A Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 134191AP150UL
Description
Glycogen Synthase Kinase-3 differs from most serine/threonine kinases in that it is active in the absence of the action of signaling pathways. Two isoforms exist, GSK-3alpha and GSK-3beta. The function of GSK-3 is to phosphorylate glycogen synthase and thereby inactivate it. Insulin action stimulates the PI 3-kinase pathway, resulting in Akt activation, which phosphorylates and inactivates GSK-3.Specifications
| GSK3A | |
| Polyclonal | |
| Unconjugated | |
| GSK3A | |
| GSK3 alpha, GSK3A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2931, 30664, 50686, 606496 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P18265, P49840, Q2NL51 | |
| Gsk3a, gsk3ab | |
| GSK3A Fusion Protein Ag4168 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat, Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction