Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GSK583, 95%

Catalog No. AC468211000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468211000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC468211000 Supplier Thermo Scientific™ Supplier No. 468211000
Add to Cart

Specifications

Chemical Name or Material GSK583
CAS 1346547-00-9
Synonym 6-(tert-Butylsulfonyl)-N-(5-fluoro-1 H-indazol-3-yl)quinolin-4-amine
Quantity 100 mg
Molecular Formula C20 H19 F N4 O2 S
Formula Weight 398.45
Physical Form Solid
Product Type Reagent
Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.