Learn More
Apexbio Technology LLC GSK621 1346607-05-3 10mg
SDP
Supplier:  Apexbio Technology LLC B602010
Encompass_Preferred
GSK621 (CAS 1346607-05-3) is a selective activator of AMP-activated protein kinase (AMPK) a serine/threonine kinase that functions as a cellular energy sensor regulating diverse metabolic pathways In cell-based assays GSK621 activates AMPK with IC50 values between 13 and 30 M Activation of AMPK by GSK621 leads to phosphorylation and inactivation of acetyl-CoA carboxylase (ACC) suppression of fatty acid synthesis inhibition of mTORC1-mediated protein synthesis and promotion of processes such as autophagy fatty acid oxidation and glucose uptake In acute myeloid leukemia (AML) cell lines and xenograft models GSK621 enhances phosphorylation of AMPK substrates and demonstrates antileukemic effects supporting its utility in studies of cellular metabolism and cancer biology
Catalog No. 50-000-90286
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.