Learn More
Medchemexpress LLC HY-104021 5mg Medchemexpress, GSK840 CAS:2361146-30-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1040215MG
Encompass_Preferred
Medchemexpress, HY-104021 5mg GSK840 CAS:2361146-30-5 GSK840 (GSK'840) is a receptor-interacting protein kinase 3 (RIP3 or RIPK3) inhibitor, which binds RIP3 kinase domain with an IC 50 of 0.9 nM, and inhibits kinase activity with an IC 50 of 0.3 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2254
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.