Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSPT1/2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143976
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143976 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143976 Supplier Invitrogen™ Supplier No. PA5143976
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human K562 whole cell, human Raji whole cell, rat pancreas tissue, mouse pancreas tissue, mouse NIH/3T3 whole cell. IHC: human gall bladder adenosquamous carcinoma tissue, human gastric adenocarcinoma tissue, human liver cancer tissue. ICC/IF: SiHa cell. Flow: JK cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Eukaryotic peptide chain release factor GTP-binding subunit ERF3A/B is an enzyme that in humans is encoded by the GSPT1,2 gene. This gene encodes a GTPase that belongs to the GTP-binding elongation factor family. The encoded protein is a polypeptide release factor that complexes with eukaryotic peptide chain release factor 1 to mediate translation termination. This protein may also be involved in mRNA stability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSPT1/2
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene GSPT2
Gene Accession No. P15170, Q149F3, Q8IYD1, Q8R050
Gene Alias ERF3A; ERF3B
Gene Symbols GSPT1, GSPT2
Host Species Rabbit
Immunogen E.coli-derived human GSPT1,2 recombinant protein (Position: K138-D499).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14852, 14853, 23708, 24420, 2935, 501582
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.