Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSPT1 Recombinant Rabbit Monoclonal Antibody (HL1345)
GREENER_CHOICE

Catalog No. PIMA547005 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547005 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547005 Supplier Invitrogen™ Supplier No. MA547005
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

eRF3a (eukaryotic peptide chain release factor subunit 3a), also known as GSPT1 (G1 to S phase transition 1), is a 499 amino acid protein that belongs to the GTP-binding elongation factor family and is involved in the regulation of cell growth, specifically via control of translation termination. Human eRF3a shares 94% sequence identity with its mouse counterpart, suggesting a conserved function between species. The gene encoding eRF3a maps to human chromosome 16, which encodes over 900 genes and comprises nearly 3% of the human genome. The GAN gene is located on chromosome 16 and, with mutation, may lead to giant axonal neuropathy, a nervous system disorder characterized by increasing malfunction with growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSPT1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1345
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene GSPT1
Gene Accession No. P15170, Q8R050
Gene Alias 551G9.2; AI314175; AI326449; AV307676; C79774; ELF; ERF3A; ETF3A; Eukaryotic peptide chain release factor GTP-binding subunit ERF3A; eukaryotic peptide chain release factor subunit 3a; eukaryotic polypeptide chain release factor 3; G1 to phase transition 1; G1 to S phase transition 1; G1 to S phase transition protein 1; G1 to S phase transition protein 1 homolog; G1st; G1-to-S phase transition 1; G1-to-S transition GTP binding protein; GSPT1; GST1; Gst-1; guanine nucleotide regulatory protein
Gene Symbols GSPT1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GSPT1. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14852, 2935, 479846
Target Species Human, Mouse, Canine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.