Learn More
Description
Immunogen sequence: AIAEALAAPS RFVLKPQREG GGNNLYGEEM VQALKQLKDS EERASYILME KIEPEPFENC LLRPGSPARV VQCISELGIF GVYVRQEKTL VMNKHVGHLL R.
Specifications
Specifications
| Antigen | GSS |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 2S9W7 |
| Concentration | 0.33 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | GSS |
| Gene Accession No. | P46413, P48637, P51855 |
| Gene Alias | AI314904; Glutathione synthase; glutathione synthetase; GS-A/GS-B; GSH S; GSH synthetase; GSHB; GSHS; GSH-S; GSS; HEL-S-64p; HEL-S-88n; MGC14098; OTTHUMP00000030711 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
