Learn More
Cytiva GST MultiTrap™ 4B Plates
Catalog No. 45001436 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
4 x 96-well Filter Plates
Catalog No. Quantity
45-001-436 4 x 96-well Filter Plates
1 options
Catalog No. 45-001-436 Supplier Cytiva Supplier No. 28405500
Add to Cart
Add to Cart
96-well filter plates for high-throughput screening and parallel purification of GST-tagged proteins
  • Highly reproducible well-to-well and plate-to-plate results using manual or robotic systems
  • High purity in one step with high chemical stability
  • Allows loading of unclarified cell lysate samples directly into wells
  • Prepacked with Glutathione Sepharose media
TRUSTED_SUSTAINABILITY
Specifications
Description GST MultiTrap FF and GST MultiTrap 4B are prepacked, 96-well filter plates for reproducible, high-throughput screening and rapid parallel purification of glutathione S-transferase (GST) tagged proteins.
Material Glutathione Sepharose 4B
Content And Storage 4°C to 30°C, 20% Ethanol
Quantity 4 x 96-well Filter Plates
Type Filter Plate