Learn More
Invitrogen™ GST Recombinant Mouse Monoclonal Antibody (GST 3-4C), HRP, Invitrogen™
Description
GST Tag recombinant mouse monoclonal antibody conjugated with HRP specifically recognizes GST Tag at both C and N-terminus of proteins. Recombinant mouse monoclonal antibodies are produced using in vitro expression systems. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Specifications
Specifications
| Antigen | GST |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | GST 3-4C |
| Concentration | 1.0 mg/mL |
| Conjugate | HRP |
| Formulation | PBS with proprietary stabilizer and proprietary preservative; pH 7.2-7.4 |
| Gene Alias | activator protein 1; AP1; AP-1; AT1G19550; cancer/testis antigen 48; c-Jun; CT48; deafness, X-linked 7; DFN7; enhancer-binding protein AP1; epididymis secretory protein Li 22; F18O14.31; F18O14_31; FAEES3; fatty acid ethyl ester synthase III; glutathione S-alkyltransferase; glutathione S-aralkyltransferase; glutathione S-aryltransferase; glutathione S-transferase M1; glutathione S-transferase mu 1; Glutathione S-transferase P; glutathione S-transferase pi 1; Glutathione-S-Transferase; gonad-specific transporter; GST; GST class-mu 1; GST class-pi; GST HB subunit 4; GST protein; GST Tag; GST1; GST3; GSTM1; GSTM1-1; GSTM1a-1a; GSTM1b-1b; GSTP; GSTP1; GSTP1-1; GTH4; GTM1; H-B; HB subunit 4; HEL-S-22; JUN; Jun activation domain binding protein; jun oncogene; jun proto-oncogene; Jun proto-oncogene, AP-1 transcription factor subunit; MU; MU-1; OATP6A1; OATP-I; OATPY; Organic anion-transporting polypeptide 6A1; Organic anion-transporting polypeptide I; p39; PI; Proto-oncogene c-Jun; S-(hydroxyalkyl)glutathione lyase; SLC21A19; SLCO6A1; Solute carrier family 21 member 19; solute carrier organic anion transporter family member 6A1; solute carrier organic anion transporter family, member 6A1; testicular tissue protein Li 174; testis-specific organic anion transporter; transcription factor AP-1; V-jun avian sarcoma virus 17 oncogene homolog; v-jun sarcoma virus 17 oncogene homolog |
| Host Species | Mouse |
| Immunogen | GST protein from Schistosoma japonicum expressed from the pGEX vector. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.