Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva GST SpinTrap™

Catalog No. 45011679 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
50 x 50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-011-679 50 x 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-011-679 Supplier Cytiva Supplier No. 28952359
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For rapid small-scale purification of GST-tagged proteins directly from clarified or unclarified cell lysates

  • Preserves protein antigenicity and function
  • Yields up to 500μg of protein per column
  • Bed volume: 50μL
  • Binding capacity/column: up to 500μg recombinant glutathione S-transferase (rGST)
  • Ligand: glutathione
  • Average particle size: 90μm
  • Binding capacity/mL chromatography medium: >25mg horse liver GST/mL medium
  • Matrix: 4% agarose
  • pH working range: 4 to 13

TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 50 mL
Binding Capacity Up to 500 μg recombinant glutathione S-transferase (rGST)
Description GST SpinTrap columns are designed for rapid small-scale purification of GST-tagged proteins directly from clarified or unclarified cell lysates
pH Range 4 to 13
Support Type 4% Agarose
Content And Storage 4 to 30°C, PBS and 0,05% Kathon CG/ICP Biocide
Particle Size 90 μm
Quantity 50 x 50 μL
For Use With (Application) For simple, small-scale purification of GST-tagged proteins and rapid expression screening by centrifugation
Type Spin Column
Ligand Type Glutathione
Sufficient For 50 Columns
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.