Learn More
Ubpbio GST-USP11 ENZYME 25 UG
Supplier:  Ubpbio H6110
Encompass_Preferred
Deubiquitinating enzymes (DUBs) are proteases that posses the ability to cleave ubiquitin chains or the isopeptide bond that conjugates ubiquitin with a substrate. Human cells have approximately 100 DUBs that play important roles in regulating various cellular events. Usp11 belongs to the ubiquitin-specific protease family. It helps in repairing DNA damage. It has been found to interact with RanBPM, a RanGTP-binding protein.UHX1
Catalog No. NC2544381
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.