Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva GSTPrep™ FF 16/10 Column

Catalog No. 45002074 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-074 1 x 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-074 Supplier Cytiva Supplier No. 28936550
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fast and easy one-step, scale-up purification of GST-tagged recombinant proteins

Glutathione Sepharose™ 4 Fast Flow is prepacked in 20mL GSTPrep columns for purification of GST-tagged proteins (as well as glutathione S-transferases and glutathione-dependent proteins). Mild elution conditions preserve protein antigenicity and function.

Features:

  • Binds approx. 200mg recombinant GST/column
  • Simple operation with pump or chromatography system (e.g., AKTA™)

GSTPrep FF 16/10 Prepacked Columns

  • Volume: 20mL
  • Dimensions: 1.6 x 10cm
  • Recommended flowrate (H2O at room temp): 1 to 10mL/min. (30 to 300cm/hr.)
  • Max. flowrate (H2O at room temp): 10mL/min. (300cm/hr.)
  • Max. pressure during operation (H2O at room temp): 1.5bar (0.15MPa, 22psi)
  • Hardware pressure limit: 5bar (0.5MPa, 73psi)
  • Storage/temperature: 20% ethanol/4° to 30°C

TRUSTED_SUSTAINABILITY

Specifications

Buffer 1M Acetate, pH 6, 0.1M NaOH, 70% Ethanol or 6M Guanidine Hydrochloride
Product Type GSTPrep Glutathione Sepharose 4 FF column
Bed Volume 20mL
Diameter (English) Inner 0.62 in.
Diameter (Metric) Inner 16mm
Quantity 1 x 20 mL
Height (English) 3.9 in.
Height (Metric) 100mm
Flow Rate <10mL/min.
Ligand Type Glutathione and 10 Carbon linker arm
Max. Pressure 1.5 bar
pH Range 3 to 12
Particle Size 90μm
Support Type 4% Highly Cross-linked Agarose
Content And Storage 4° to 30°C
Column Type Ion Exchange Column
For Use With (Application) Purification of Glutathione S-transferases and Glutathione-dependent Proteins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.