Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva GSTrap™ FF Columns: 7 x 25mm

Catalog No. 45000282 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
2 x 1 mL
5 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-282 2 x 1 mL
45-000-281 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-000-282 Supplier Cytiva Supplier No. 17513002
Add to Cart
Add to Cart

Purifies GST-tagged proteins in one step

  • Mild elution conditions preserve antigenicity and function of proteins extracted
  • Prepacked with Glutathione Sepharose™ media
  • Average particle size: 90μm
  • Bed dimensions: 7 × 25mm
  • Flow velocity: <4mL/min.
Media
  • Matrix: Highly cross-linked 4% agarose
  • Ligand: glutathione and 10-carbon linker arm
  • Binding capacity: >10mg/mL
  • pH stability working range: 3 to 12

TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 1 mL
Diameter (English) Inner 0.27 in.
Diameter (Metric) Inner 7 mm
Description GSTrap FF columns are prepacked Glutathione Sepharose Fast Flow columns for fast, convenient, one-step purification of glutathione S-transferase (GST) tagged proteins.
Flow Rate <4 mL/min.
Material Polypropylene
pH Range 3 to 12
Support Type 4% Highly Cross-linked Agarose
Content And Storage 4°C to 30°C, 20% Ethanol
Max. Operating Pressure 5 bar (0.5 MPa, 70 psi)
Particle Size 90 μm
Quantity 2 x 1 mL
For Use With (Application) For the purification of GST-tagged proteins from bacterial lysates and other glutathione S-tranferases or glutathione-dependent proteins and for preparation of cytoplasmic extract
Type Pre-packed Column
Ligand Type Glutathione and 10 carbon linker arm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.