Learn More
Abcam Goat Anti-Rabbit IgG H&L (APC) preadsorbed. Reacts with the heavy and light chains of Rabbit IgG, and with the light chains of Rabbit IgM. Goat Anti-Rabbit IgG H&L (Allophycocyanin) preadsorbed shows minimal cross reactivity with Human and Mouse immunoglobulins.
Supplier:  Abcam AB130805125UG
Encompass_Preferred
Goat Anti-Rabbit IgG H&L (APC) preadsorbed. Reacts with the heavy and light chains of Rabbit IgG, and with the light chains of Rabbit IgM. Goat Anti-Rabbit IgG H&L (Allophycocyanin) preadsorbed shows minimal cross reactivity with Human and Mouse immunoglobulins.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1757372
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.