Learn More
Biocare Medical, LLC 4plus Biotinylated Goat Anti-Mouse IgG - 25 ml
SDP
Supplier:  Biocare Medical, LLC GM601H
Encompass_Preferred
This biotinylated antibody reacts with heavy and light chains on mouse IgG. It is adsorbed against horse, bovine and human. It reacts with all mouse IgG subclasses and mouse IgM. It has been affinity-purified with minimal cross-reaction to human, bovine and horse serum proteins. A long spacer arm has been positioned between the biotin and the secondary antibody, which provides a significant increase in sensitivity when coupled with streptavidin conjugates. Biocare Goat Anti-Mouse IgG can be used with either streptavidin horseradish peroxidase or streptavidin alkaline phosphatase detection systems.
Catalog No. 50-828-65
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.