Learn More
Invitrogen™ GTF2B Monoclonal Antibody (IIB8)

Description
MA1-10884 detects Transcription Factor IIB from human and mouse samples. MA1-10884 has been successfully used in ELISA, Western blot, and immunoprecipitation applications. The MA1-10884 immunogen is recombinant human transcription factor IIB (TFIIB).
Specifications
Specifications
| Antigen | GTF2B |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | IIB8 |
| Concentration | ∼1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | GTF2B |
| Gene Accession No. | P62915, Q00403 |
| Gene Alias | general transcription factor 2B L homeolog; general transcription factor IIB; general transcription factor TFIIB; GTF2B; gtf2b.L; gtf2b-a; gtf2b-b; LOW QUALITY PROTEIN: transcription initiation factor IIB; MGC137096 protein; RNA polymerase II alpha initiation factor; RNA polymerase II transcription factor IIB; RP11-82K18.1; S300-II; TF2B; tfiib; Transcription initiation factor IIB; Transcription initiation factor IIB-like protein; wu:fc44c11; XELAEV_18023356mg; XLTFIIB; zgc:66258 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.