Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF2E2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555220
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555220 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555220 Supplier Invitrogen™ Supplier No. PA555220
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSLLRERELF KKRALSTPVV EKRSASSESS SSSSKKKKTK VEHGGSSGSK QNSDHSNGSF NLKALSGSSG YKFGVL Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

The general transcription factor IIE (TFIIE) is part of the RNA polymerase II transcription initiation complex, recruiting TFIIH and being essential for promoter clearance by RNA polymerase II. TFIIE is a heterodimer (and sometimes heterotetramer) of alpha and beta subunits. The protein encoded by this gene represents the beta subunit of TFIIE.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF2E2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GTF2E2
Gene Accession No. P29084, Q9D902
Gene Alias 34kDa; AI462509; C330006J08Rik; FE; general transcription factor II E, polypeptide 2 (beta subunit); general transcription factor II E, polypeptide 2 (beta subunit, 34kD); general transcription factor IIE subunit 2; general transcription factor IIE, polypeptide 2 (beta subunit); general transcription factor IIE, polypeptide 2 (beta subunit, 34kD); general transcription factor IIE, polypeptide 2, beta; general transcription factor IIE, polypeptide 2, beta 34kDa; Gtf2e2; T2EB; TF2E2; TFIIE beta subunit; TFIIE-B; TFIIE-beta; transcription initiation factor IIE subunit beta; TTD6
Gene Symbols GTF2E2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GTF2E2. Recombinant protein control fragment (Product #RP-93951).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2961, 306516, 68153
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.