Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTF2E2/TF2E2/TFIIE-beta Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501569406
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-9406 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-9406 Supplier Bethyl Laboratories, Inc Supplier No. A305054AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Recruits TFIIH to the initiation complex and stimulates the RNA polymerase II C-terminal domain kinase and DNA-dependent ATPase activities of TFIIH. Both TFIIH and TFIIE are required for promoter clearance by RNA polymerase. Peterson M.G., Nature 354:369-373(1991). Sumimoto H., Nature 354:401-404(1991). Kang S.-W., Genes Cells 5:251-263(2000).

Specifications

Antigen GTF2E2/TF2E2/TFIIE-beta
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene GTF2E2
Gene Accession No. P29084
Gene Alias FE, TF2E2, TFIIE-B, TFIIE-beta, general transcription factor IIE subunit 2, transcription initiation factor IIE subunit beta
Gene Symbols GTF2E2
Host Species Rabbit
Immunogen Between 241 and 291
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2961
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.