Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF2F1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109796
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109796 Supplier Invitrogen™ Supplier No. PA5109796
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSAEGGSTSS TLRAAASKLE QGKRVSEMPA AKRLRLDTGP QSLSGKSTPQ PPSGKTTPNS GDVQVTEDAV RRYLTRKPMT TKDLLKKFQT KKTGLSSEQT VNVLAQILKR LNPERKMIND KMHFSLKE.

TFIIF is a general transcription initiation factor that binds to RNA polymerase II and helps to recruit it to the initiation complex in collaboration with TFIIB. It promotes transcription elongation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF2F1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Gtf2f1
Gene Accession No. P35269, Q3THK3, Q6AY96
Gene Alias 2810405L04Rik; BTF4; C76800; General transcription factor IIF 74 kDa subunit; general transcription factor IIF polypeptide 1 (74kD subunit); general transcription factor IIF subunit 1; general transcription factor IIF, polypeptide 1; general transcription factor IIF, polypeptide 1, 74kDa; Gtf2f1; MGC94148; OTTHUMP00000237859; RAP74; TF2F1; TFIIF; TFIIF-alpha; transcription initiation factor IIF subunit alpha; Transcription initiation factor RAP74
Gene Symbols Gtf2f1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 390-517 of GTF2F1 (NP_0020872).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2962, 316123, 98053
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.