Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTF3C5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7203582 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7166 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7166 Supplier Proteintech Group Inc Supplier No. 114472AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lamina-associated polypeptide 2 (LAP2), isoform beta is a protein that in humans is encoded by the TMPO gene. LAP2 is an inner nuclear membrane (INM) protein. LAP2 plays a role in the regulation of nuclear architecture by binding lamin B1 and chromosomes. This interaction is regulated by phosphorylation during mitosis.

Specifications

Antigen GTF3C5
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene GTF3C5
Gene Accession No. Q8R2T8, Q9Y5Q8
Gene Alias GTF3C5, TF3C epsilon, TFIIIC 63 kDa subunit, TFiiiC2 63, TFIIIC63, TFIIICepsilon
Gene Symbols Gtf3c5
Host Species Rabbit
Immunogen GTF3C5 Fusion Protein Ag1999
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362095, 70239, 9328
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.