Learn More
GTF3C5/TFIIIC63 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301242A
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
GTF3C5 is polypeptide 5 of general transcription factor IIIon Channel. Human transcription factor IIIon Channel (hTFIIIon Channel) is a multisubunit complex that directly recognizes promoter elements and recruits TFIIIB and RNA polymerase III.Specifications
| GTF3C5/TFIIIC63 | |
| Polyclonal | |
| Unconjugated | |
| GTF3C5 | |
| general transcription factor IIIC, polypeptide 5 (63kD), general transcription factor IIIC, polypeptide 5, 63kDa, TF3C-epsilon, TFIIIC 63 kDa subunit, TFiiiC2-63, TFIIIC63, TFIIICepsilon, transcription factor IIIC 63 kDa subunit, transcription factor IIIC subunit epsilon, transcription factor IIIC, 63 kD | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 70239, 9328 | |
| 4° C | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q8R2T8, Q9Y5Q8 | |
| Gtf3c5 | |
| Between 469 and 519 | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.