Learn More
Apexbio Technology LLC GTP-Binding Protein Fragment, G alpha 5mg
SDP
Supplier:  Apexbio Technology LLC A10225
Encompass_Preferred
GTP-Binding Protein Fragment G alpha is a peptide fragment derived from the amino-terminal region of the alpha subunit of GTP-binding proteins It is designed to aid in elucidating the mechanisms of membrane association and -subunit interactions in cell signaling pathways GTP-Binding Protein Fragment G alpha exerts its biological activity by retaining membrane-binding properties independent of the nucleotide-binding state anchoring the protein to the cytoplasmic membrane surface This peptide fragment serves as an experimental tool to study membrane-association mechanisms and signal transduction pathways in biochemical and biomedical research Based on these properties GTP-Binding Protein Fragment G alpha holds research potential in investigating the molecular basis of G protein-mediated signal transduction and protein-membrane interactions
Catalog No. 50-000-95635
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.