Learn More
                                    CPC Scientific H-Cys-Lys-Gln-Leu-Gln-Lys-Asp-Lys-Gln-Val-Tyr-Arg-Ala-Thr-His-Arg-OH (trifluoroacetate salt) 0.5MG
Supplier: CPC Scientific GHRF011A
            ONE-LETTER SEQUENCE: CKQLQKDKQVYRATHRMOLECULAR FORMULA: C85H144N30O24S1MOLECULAR WEIGHT:2002.23STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [101038-78-2], SYNONYMS: [Cys0]-GTP-Binding Protein Gs alpha (28-42)RESEARCH AREA: HormonalREFERENCES: S.M. Mumby et al., PNAS, 83, 265 (1986)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.