Learn More
Promega GTPase-Glo Assay, 1,000 assays
Catalog No. PRV7681 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1000 Assays
Catalog No. Quantity
PRV7681 1000 Assays
1 options
Catalog No. PRV7681 Supplier Promega Supplier No. V7681
The GTPase-Glo Assay assesses the activities of GTPases, GAPs and GEFs, which are components of the GTPase cycle, by detecting the amount of GTP remaining after GTP hydrolysis in a GTPase reaction.
  • Simple, Sensitive GTPase Activity Assay
  • Simple add−and−read format
  • Suitable for 96−, 384− and 1536−well plates
  • Low background and large dynamic range
Specifications
Quantity 1000 Assays