Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanidine-HCl, Molecular Grade, Promega™

Catalog No. PRH5381 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-H5381 100 g
PR-H5383 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PR-H5381 Supplier Promega Corporation Supplier No. H5381
Only null left
Add to Cart
Add to Cart

Used for the isolation of intact mRNA from tissues or cultured cells.

  • Quality tested and certified free of DNase, RNase and protease activity

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735

Specifications

Color Colorless or White
Quantity 100 g
Formula Weight 95.53
Grade Molecular
Physical Form Fine Crystals
Chemical Name or Material Guanidine-HCl, For the isolation of intact mRNA from tissues or cultured cells
Recommended Storage Store at 22° to 25°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.