Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Guanidine thiocyanate, 99%, for biochemistry

Catalog No. AC411110010
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC411110010 1 kg Plastic bottle
AC411111000 100 g Plastic bottle
AC411112500 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC411110010 Supplier Thermo Scientific Chemicals Supplier No. 411110010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 593-84-0 | CH5N3·CHNS | 118.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Guanidine thiocyanate is a useful tool for RNA isolation and protein solubilization. It is involved in the quantification of mRNA from hepatocytes. It acts as a medium for blood smaple lystae in order to maintain the integrity of nucleic acid. It is a chaotropic agent, mainly involved in the extraction of DNA and RNA. It is utilized to deactivate the influenza virus.

Solubility
Soluble in water and ethanol

Notes
Light sensitive. Incompatible with acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-84-0
Molecular Formula CH5N3·CHNS
Molecular Weight (g/mol) 118.16
MDL Number MFCD00013027

Specifications

Absorbance (3M soln. in water), 0.05 max. at 300nm, (3 M soln. in water), (3 M soln. in water) at 0nm
Melting Point 118°C to 122°C
Color White or Colorless
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Plastic bottle
Linear Formula NH2C(NH)NH2·HSCN
Quantity 1 kg
UN Number 3261
Solubility Information Solubility in water: 1420g/L (20°C)
Vapor Pressure Negligible
Formula Weight 118.16
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Guanidine thiocyanate, 0.99
Show More Show Less
Hazard Category Skin corrosion/irritation (category 1), Hazardous to the aquatic environment (chronic category 3), Acute toxicity (category 4)
Hazard Statement Corrosive to the respiratory tract, Causes severe skin burns and eye damage, Contact with acids liberates very toxic gas, Harmful to aquatic life with long lasting effects, Harmful if swallowed, in contact with skin or if inhaled
EINECSNumber 209-812-1
RTECSNumber XL1225000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.