accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NH2HNHSN

CAS RN 593-84-0

IUPAC Name: guanidine; sulfanylformonitrile
Synonyms: guanidinium thiocyanate
Molecular Weight (g/mol): 118.16
Molecular Formula: C2H6N4S
InChi Key: ZJYYHGLJYGJLLN-UHFFFAOYSA-N
SMILES: SC#N.NC(N)=N
Quantity 
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (4)
Physical Form 
  • (1)
  • (5)
  • (6)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
Grade 
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (6)
  • (1)
  • (4)
  • (6)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (5)
  • (6)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)