Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanidine Thiocyanate, MP Biomedicals™

Catalog No. MP04820991 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP04820991 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP04820991 Supplier MP Biomedicals, Inc Supplier No. 04820991
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Guanidine Thiocyanate, Research Grade, >98%, Hard, White Crystalline Granules

  • Guanidine Thiocyanate is often used in the isolation of intact ribonucleic acid to eliminate RNase activity.
  • A 4 M solution of guanidine thiocyanate irreversibly inactivates RNase.
  • In the presence of guanidine thiocyanate, proteins dissolve readily, cellular structures disintegrate and nucleoproteins dissociate from nucleic acids, as protein secondary structure is lost.
  • Guanidine Thiocyanate is a chaotropic agent and strong protein denaturant which inactivates nucleases approximately 2.5 times faster than guanidine hydrochloride.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-84-0
Molecular Formula C2H6N4S
Molecular Weight (g/mol) 118.16
MDL Number MFCD00013027
InChI Key ZJYYHGLJYGJLLN-UHFFFAOYSA-N
Synonym Guanidinium rhodanide, Guanidinium thiocyanate

Specifications

Melting Point 115°C to 122°C
Color White
pH 5 to 7 (6M aq soln)
Loss on Drying <1%
Quantity 500 g
UN Number UN1759
Beilstein 3563461
Solubility Information Soluble in (6.0 M aq soln): Clear, Colorless Solution.
Grade >98%
Physical Form Crystalline Granules
storageNote1 Store at Room Temperature (15-30°C). Protect from light.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.