Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanine, 98%, MP Biomedicals™

Catalog No. ICN19069180 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19069180 100 g
ICN19069101 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19069180 Supplier MP Biomedicals, Inc Supplier No. 0219069180
Add to Cart
Add to Cart

Guanine is one of the four main nucleobases found in the nucleic acids DNA and RNA

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-40-5
Molecular Formula C5H5N5O
Molecular Weight (g/mol) 151.13
MDL Number MFCD00071533
InChI Key UYTPUPDQBNUYGX-UHFFFAOYSA-N
Synonym guanine, 2-amino-6-hydroxypurine, guanin, 2-aminohypoxanthine, mearlmaid, pearl essence, guanine enol, stella polaris, dew pearl, natural pearl essence
PubChem CID 764
ChEBI CHEBI:16235
IUPAC Name 2-amino-6,7-dihydro-3H-purin-6-one
SMILES NC1=NC(=O)C2=C(N1)N=CN2

Specifications

Color White
Quantity 100 g
Formula Weight 151.13
Physical Form Powder
Chemical Name or Material Guanine
EINECSNumber 200-799-8
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.