Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Guanine Nucleobase, >99% Purity, Powder, 25 g

Catalog No. 502704806
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4806 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4806 Supplier Abcam Supplier No. AB14624225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Purine derivative and nucleobase for incorporation into RNA and DNA with >99% purity.

  • Molecular Weight: 151.13 g/mol
  • Purity: >99%
  • Form: Powder
  • Storage Buffer: 100% Guanine
  • Storage Conditions: Store at Ambient.
  • Shipping Conditions: Ambient - Can Ship with Ice
  • Description: Nucleobase incorporated into nucleic acids, such as RNA and DNA; purine derivative.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at ambient temperature.
Shipping Condition Ambient
Form Powder
Quantity 25 g
For Use With (Application) Sample Prep and Detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.