Learn More
Abcam Guanosine 3',5'-cyclic monophosphate sodium salt (cGMP), cyclic nucleotide second messenger, 500MG
Supplier:  Abcam AB120805500MG
Encompass_Preferred
MW 367.19 Da, Purity >99%. Cyclic nucleotide second messenger derived from guanosine triphosphate. Generated by guanylate cyclases. Involved in a number of cellular processes; downstream targets include cyclic nucleotide gated channels, cGMP-regulated phosphodiesterases and cGMP-dependent kinases.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7248
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.