Learn More
Abcam Guanosine 5'-Diphosphate Disodium Salt, >98% Purity, 500 mg
Description
- Molecular Weight: 487.17 g/mol
- Purity: >98%
- Applications: Substrate of pyruvate kinase to produce GTP in support of RNA biosynthesis; used to study the kinetics and characteristics of GTPases (e.g., GPCRs) and cell signaling processes mediated by guanine nucleotide exchange factors
- Chemical Composition: 100% Guanosine-5'-diphosphate [GDP] disodium salt
- CAS Number: 7415-69-2
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Guanosine 5'-diphosphate |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Substrate | Pyruvate kinase |
| Content And Storage | Store at -20°C. |
| Form | Powder |
| Buffer | 100% Guanosine-5'-diphosphate [GDP] disodium salt |
| Formulation | 100% Guanosine-5'-diphosphate [GDP] disodium salt |
| Quantity | 500 mg |
| For Use With (Application) | RNA biosynthesis, GTPase kinetics, GPCR studies, cell signaling studies |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.