Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanosine-5'-triphosphate Disodium Salt, ≥95%, MP Biomedicals™

Catalog No. ICN19521325 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
250 mg
500 mg
1 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19521325 25 mg
ICN19521310 10 mg
02-195-21380 100 mg
ICN19521383 250 mg
ICN19521390 500 mg
ICN19521391 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. ICN19521325 Supplier MP Biomedicals, Inc Supplier No. 0219521325
Add to Cart
Add to Cart

Guanosine-5'-triphosphate Disodium Salt, ≥95% is a substrate for RNA polymerases.

  • Guanosine 5′-triphosphate (GTP), a purine trinucleotide, is a substrate for RNA polymerases and a wide range of GTPase including G-protein coupled receptor (GPCR) GTPases and cell signaling and cycling associated guanine nucleotide exchange factors (GEF).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36051-31-7
Molecular Formula C10H16MgN5O14P3
Molecular Weight (g/mol) 547.49
MDL Number MFCD03410297
InChI Key NMQPZZLDIUETGE-GWTDSMLYSA-N
Synonym guanosine 5'-triphosphate, guanosine-5'-triphosphate disodium salt dihydrate gtp, guanosine-triphosphate, guanosine 5'-triphosphate 4-, gtp 4-, 2r,3s,4r,5r-5-2-amino-6-hydroxy-9h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxy phosphonatooxy phosphinate, 2r,3s,4r,5r-5-2-amino-6-oxo-3h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methoxy-oxidophosphoryl oxy-oxidophosphoryl phosphate
PubChem CID 131676145
SMILES [Mg].NC1=NC(=O)C2=C(N1)N(C=N2)[C@@H]1O[C@H](COP(O)(=O)OP(O)(=O)OP(O)(O)=O)[C@@H](O)[C@H]1O

Specifications

Absorbance λ max (water): 252 ± 3 nm
Color White
Assay Percent Range ≥95%
Quantity 25 mg
IUPAC Name ({[({[(2R,3S,4R,5R)-5-(2-amino-6-oxo-6,9-dihydro-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy](hydroxy)phosphoryl}oxy)phosphonic acid magnesium
Formula Weight 567.2
Percent Purity ≥95%
Physical Form Powder
Chemical Name or Material Guanosine-5'-triphosphate disodium salt, For HPLC analysis
EINECSNumber 259-940-7
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.