Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Guanosine 5'-Triphosphate Trisodium Salt, >90% Purity, 100 mg

Catalog No. 502583228
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3228 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3228 Supplier Abcam Supplier No. AB146528100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nucleoside triphosphate that activates signal transducing G proteins and serves as an energy-rich precursor for DNA and RNA biosynthesis.

  • Molecular weight: 589.13 g/mol
  • Purity: >90%
  • Function: Nucleoside triphosphate; carrier of phosphates and pyrophosphates for biosynthetic pathways; activator of signal transducing G proteins; precursor for DNA and RNA biosynthesis
  • Storage temperature: Store at -20°C
  • Shipping conditions: Ambient - Can Ship with Ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Form Powder
Shipping Condition Ambient
Content And Storage Store at -20°C.
Target Molecule G proteins
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.