Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Guanosine 5'-Triphosphate Trisodium Salt, >90% Purity, 500 mg

Catalog No. 502583227
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3227 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3227 Supplier Abcam Supplier No. AB146528500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Activates signal transducing G proteins and functions as a phosphate carrier in biosynthetic pathways.

  • Molecular Weight: 589.13 g/mol
  • Purity: >90%
  • Storage Conditions: Store at -20°C
  • Shipping Conditions: Ambient - Can Ship with Ice
  • Biological Function: Nucleoside triphosphate; functions as a carrier of phosphates and pyrophosphates for biosynthetic pathways; activates signal transducing G proteins; precursor for DNA and RNA biosynthesis

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule G proteins
Shipping Condition Ambient
Content And Storage Store at -20°C.
Quantity 500 mg
For Use With (Application) Activation of signal transducing G proteins, DNA and RNA biosynthesis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.