Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanylate kinase Antibody (OTI4A8) Alexa Fluor™ Plus 594, R&D Systems™
SDP

Catalog No. RD10070057
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10070057 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10070057 Supplier R&D Systems Supplier No. NBP270869AFP594

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

Guanylate kinase Monoclonal antibody specifically detects Guanylate kinase in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),CyTOF

Specifications

Antigen Guanylate kinase
Applications Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone OTI4A8
Conjugate Alexa Fluor Plus 594
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.4.8, FLJ42686, FLJ43710, GMK, GMP kinase, guanylate kinase, guanylate kinase 1
Host Species Mouse
Immunogen Full length human recombinant protein of human GUK1 (NP_000849) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2987
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.