Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanylthiourea 98.0+%, TCI America™
SDP

Catalog No. G0234100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0234100G 100 g
G023425G 25 g
G0234500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G0234100G Supplier TCI America Supplier No. G0234100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2114-02-5
Molecular Formula C2H6N4S
Molecular Weight (g/mol) 118.16
MDL Number MFCD00014472
InChI Key OKGXJRGLYVRVNE-UHFFFAOYSA-N
Synonym amidinothiourea, guanylthiourea, guthimin, guthimine, 2-imino-4-thiobiuret, n-amidinothiourea, thiourea, aminoiminomethyl, thiodicyanodiamidine, diaminomethylidene thiourea, gutimine
PubChem CID 2724563
IUPAC Name diaminomethylidenethiourea
SMILES C(=NC(=S)N)(N)N

Specifications

Color White
Quantity 100 g
Formula Weight 118.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Guanylthiourea
EINECSNumber (2)-1780
RTECSNumber YR8250000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.