Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanylurea sulfate, 97%

Catalog No. p-3758136
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC411140250 25 g Plastic bottle
AC411141000 100 g Plastic bottle
AC411145000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC411140250 Supplier Thermo Scientific Chemicals Supplier No. 411140250
Only null left
Add to Cart

CAS: 591-01-5 | C4H12N8O2·H2SO4 | 302.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-01-5
Molecular Formula C4H12N8O2·H2SO4
Molecular Weight (g/mol) 302.26
MDL Number MFCD00035565

Specifications

Melting Point 190.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula [NH2C(NH)NHCONH2]2·H2SO4
Quantity 25 g
Merck Index 15,3104
Solubility Information Solubility in water: soluble.
Formula Weight 302.26
Percent Purity 98%
Physical Form Fine Powder
Chemical Name or Material Guanylurea sulfate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 209-697-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.