Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Guar Gum
SCI_ED

Catalog No. s25346 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Solid
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25346 100 g Solid
S25346A 500 g Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25346
Used in slime-making experiments

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 9000-30-0
Molecular Formula C10H14N5Na2O12P3
Molecular Weight (g/mol) 535.146
InChI Key JEKDCIBJADJZSK-UHFFFAOYSA-L
Synonym guar gum
PubChem CID 44134661
IUPAC Name disodium;[[[5-(6-aminopurin-9-yl)-3-hydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-oxidophosphoryl] hydrogen phosphate
SMILES C1C(C(OC1N2C=NC3=C2N=CN=C3N)COP(=O)(O)OP(=O)([O-])OP(=O)(O)[O-])O.[Na+].[Na+]

Specifications

Color White-Yellow
Assay Percent Range 100% w/v
Quantity 100 g
Packaging Solid
Chemical Name or Material Guar Gum
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.