Learn More
Guardian Protective Wear Air Weave Bib-Style Rainwear Pants
Catalog No. 19182112
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-182-112 Medium
19-182-114 X-Large
19-182-111 Small
19-182-113 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-182-112 Supplier Guardian Protective Wear Supplier No. 855YM
Add to Cart
Add to Cart
This item is not returnable. View return policy
Protection gear for rain and moisture
  • Breathable polyurethane on nylon
  • Flame retardant to NFPA 701 standards
  • Dielectric and anti-static
  • Flap pocket on right hip with hook and loop closure
  • Snaps on waistband and cuff for adjustable fit
  • Zipper-fly closure
  • Color: Yellow
Specifications
Size Medium
Color Yellow