Learn More
Guardian Protective Wear Air Weave Bib-Style Rainwear Pants
Description
- Breathable polyurethane on nylon
- Flame retardant to NFPA 701 standards
- Dielectric and anti-static
- Flap pocket on right hip with hook and loop closure
- Snaps on waistband and cuff for adjustable fit
- Zipper-fly closure
- Color: Yellow
Specifications
Specifications
| Size | Large |
| Color | Yellow |