Learn More
Guardian Protective Wear Tri-Weave Overalls
Specifications
Specifications
| Closure Type | Adjustable Suspenders |
| Color | Yellow |
| Material | Nylon, Vinyl |
| Seam Type | Stitched, Taped |
| Size | Medium |
| Construction | Tri-weave |
| For Use With (Application) | Protection from chemicals |
| Style | Bib |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.