Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec™ Guardian Safe View Body Bag with Envelope Zipper, without Handles
SDP

Unique optically clear interior panel on the Guardian Safe View Body Bag allows viewing the remains contained in the bag with minimal exposure risk to personnel. Both panels are secured with envelope style, heavy-duty zippers with dual zipper pulls for easy access.

Supplier:  Mopec™ BE175

Catalog No. 22-445-816


Description

Description

  • Standard Duty
  • Made of Reinforced Coextruded Polyethylene Envelope (C-shape)
  • Heat Sealed Seams
  • Suitable for Cremation
  • Clear Viewing Panel
Specifications

Specifications

300 lb.
Black
Adult
96 x 36 in.
243.84 cm
91.44 cm
Envelope Zipper
Polyethylene
10 mil
96 in.
36 in.
Body Bag
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.