Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ GuardTec3™ Cut-Resistant Gloves with Nitrile Palm

Catalog No. 19148833 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-148-833 X-Small
19-148-834 Small
19-148-835 Medium
19-148-836 Large
19-148-837 X-Large
19-148-838 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-148-833 Supplier Wells Lamont™ Supplier No. Y9286XS

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Cut-resistant gloves provide a comfortable fit while nitrile palm reassures secure grip on dry or oily parts.

  • High cut-resistance fiber provides protection from lacerations
  • Sandy nitrile palm coating technology offers easy grip for wet and oily parts
  • Dark-colored shell conceals dirt and grime

Specifications

Color Gray
Cut Resistance Rating ANSI A3
Material Nitrile
Product Type Cut-resistant Gloves
Size X-Small
For Use With (Application) Easy grip for wet and oily parts and provides protection from lacerations and superior resistance against cuts
Glove Cuff Color Yellow
Glove Cuff Style Overcast Hem
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.