Learn More
Wells Lamont™ GuardTec3™ Cut-Resistant Gloves with Nitrile Palm
Shop All Wells Lamont Industry Group ProductsDescription
- High cut-resistance fiber provides protection from lacerations
- Sandy nitrile palm coating technology offers easy grip for wet and oily parts
- Dark-colored shell conceals dirt and grime
Specifications
Specifications
| Color | Gray |
| Cut Resistance Rating | ANSI A3 |
| Material | Nitrile |
| Product Type | Cut-resistant Gloves |
| Size | Small |
| For Use With (Application) | Easy grip for wet and oily parts and provides protection from lacerations and superior resistance against cuts |
| Glove Cuff Color | Red |
| Glove Cuff Style | Overcast Hem |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.