Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ GuardTec4 EliteGrip™ Gloves with Nitrile Palm

Catalog No. 19148841 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Glove Cuff Color:
Black
Blue
Brown
Green
Red
Yellow
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Glove Cuff Color Size
19-148-841 Green Medium
19-148-839 Yellow X-Small
19-148-840 Red Small
19-148-842 Brown Large
19-148-843 Blue X-Large
19-148-844 Black 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-148-841 Supplier Wells Lamont™ Supplier No. Y9288M
This item is not returnable. View return policy

Hand protection with dexterity and fit

  • Feature highly cut-resistant fiber that provides protection against laceration
  • Sandy nitrile palm coating provides excellent grip in wet and oily environments
  • Dark-colored shell conceals dirt and grime
  • Color: Gray

Specifications

Color Gray
Cut Resistance Rating ANSI A4
Material Nitrile
Product Type Cut-resistant Gloves
Size Medium
For Use With (Application) Good grip in wet and oily environment and provides protection from lacerations and superior resistance against cuts
Glove Cuff Color Green
Glove Cuff Style Knit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.