Learn More
Wells Lamont™ GuardTec4 EliteGrip™ Gloves with Nitrile Palm
Shop All Wells Lamont Industry Group ProductsDescription
- Feature highly cut-resistant fiber that provides protection against laceration
- Sandy nitrile palm coating provides excellent grip in wet and oily environments
- Dark-colored shell conceals dirt and grime
- Color: Gray
Specifications
Specifications
| Color | Gray |
| Cut Resistance Rating | ANSI A4 |
| Material | Nitrile |
| Product Type | Cut-resistant Gloves |
| Size | Large |
| For Use With (Application) | Good grip in wet and oily environment and provides protection from lacerations and superior resistance against cuts |
| Glove Cuff Color | Brown |
| Glove Cuff Style | Knit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.