Learn More
Description
GUCY2C Recombinant Monoclonal Antibody for Flow, ELISA
Specifications
Specifications
| Antigen | GUCY2C |
| Applications | ELISA, Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Clone | 4G12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 |
| Gene | GUCY2C |
| Gene Accession No. | P25092 |
| Gene Alias | AI893437; DIAR6; Gcc; GC-C; guanylate cyclase 2C; guanylyl cyclase C; GUC2C; GUCY2C; heat stable enterotoxin receptor; heat-stable enterotoxin receptor; hSTAR; intestinal guanylate cyclase; MECIL; MUCIL; STA receptor; STAR |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
