Learn More
Med Vet International McKesson Tourniquet, Quick Release, 14 Inch Length, 63-4418, 12/Box
Supplier:  Med Vet International 581564BX
Encompass_Preferred
McKesson Tourniquet, Quick Release 14 Inch Length Synthetic Silk and Terylene 14-inches in length with a 1-inch width, One size fits all Automatic clamp and quick release buttons allow for quick response times Made from synthetic silk and terylene material; suitable for those with latex allergies Blue in color Reusable
Catalog No. 50-304-4231
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.