Learn More
Med Vet International McKesson Perry Performance Plus Surgical Glove, Size 8, Cream, 20-1080N, 50 Pairs
Supplier: Med Vet International 1044708BX
Surgical Glove McKesson Perry Performance Plus Size 8 Sterile Latex Standard Cuff Length Smooth Cream Not Chemo Approved Size 8 Cream Length (average): 11 inch (280 mm) Fingertip thickness (single): 9 mil (0.23 mm) Sterilization: Gamma radiation Anti-slip, smooth finish Manufactured with SureFit Technology which is designed to enhance roll down protection Continuous polymer coating for excellent dry- and damp-hand donnability
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.